EMAIL THIS PAGE TO A FRIEND
32093 Sigma-Aldrich

Probenazole

PESTANAL®, analytical standard

Synonym: 3-(2-Propenyloxy)-1,2-benzisothiazole 1,1-dioxide, 3-Allyloxy-1,2-benzisothiazole 1,1-dioxide

  • CAS Number 27605-76-1

  • Empirical Formula (Hill Notation) C10H9NO3S

  • Molecular Weight 223.25

  •  Beilstein Registry Number 1214464

  •  MDL number MFCD00274602

  •  PubChem Substance ID 329753734

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Antifungals Standards,
grade   analytical standard
InChI Key   WHHIPMZEDGBUCC-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Probenazole may have been used as working standard in LC–MS/MS analysis for determination of pesticide residue from paprika extract.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

General description

Probenazole is a herbicide very effective in treating rice blast disease. It is a known active ingredient of Oryzemate.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?