• USA Home
  • 32554 - Chlorpromazin-d6 hydrochloride

EMAIL THIS PAGE TO A FRIEND
32554 Supelco

Chlorpromazin-d6 hydrochloride

VETRANAL®, analytical standard

Synonym: 2-Chloro-10-(3-dimethyl-d6-aminopropyl)phenothiazine

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antipsychotics, Chromatography,
Quality Level   100
grade   analytical standard
product line   VETRANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
Veterinary
format   neat
mass shift   M+6
SMILES string   Cl.[2H]C([2H])([2H])N(CCCN1c2ccccc2Sc3ccc(Cl)cc13)C([2H])([2H])[2H]
InChI   1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H/i1D3,2D3;
InChI key   FBSMERQALIEGJT-TXHXQZCNSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811PIH 6.1 / PGI
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?