EMAIL THIS PAGE TO A FRIEND
35376 Sigma-Aldrich

Cuelure

PESTANAL®, analytical standard

Synonym: 4-(3-Oxobutyl)phenyl acetate, 4-(4-Acetoxyphenyl)-2-butanone, Cue-lure, Q-lure

  • CAS Number 3572-06-3

  • Linear Formula CH3CO2C6H4CH2CH2COCH3

  • Molecular Weight 206.24

  •  Beilstein Registry Number 1961620

  •  EC Number 222-682-0

  •  MDL number MFCD00008704

  •  PubChem Substance ID 329755140

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, All Alphabetically Sorted, Analytical Standards,
grade   analytical standard
InChI Key   UMIKWXDGXDJQJK-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.509(lit.)
bp   123-124 °C/0.2 mmHg(lit.)
density   1.099 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
EL8950000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

香精香料分析

我们针对全部三个分析阶段提供了优质的产品解决方案,可助您快速获得精确的结果。通过以下内容,您可以详细了解我们的样品制备、分离、检测和分析工作流程解决方案。在检测纯度、定性或确证成分时,速度和分离度是关键。您可以在此搜索特定的产品或下载相关应用。
Keywords: Calorimetry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?