EMAIL THIS PAGE TO A FRIEND
35493 Sigma-Aldrich

Kelevan

PESTANAL®, analytical standard

  • CAS Number 4234-79-1

  • Empirical Formula (Hill Notation) C17H12Cl10O4

  • Molecular Weight 634.80

  •  Beilstein Registry Number 2197868

  •  MDL number MFCD00055538

  •  PubChem Substance ID 329755176

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Cyclodienes, H-M,
grade   analytical standard
InChI Key   POSKOXIJDWDKPH-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
PC8400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?