• USA Home
  • 36792 - 1,1′-Diethyl-4,4′-bipyridinium dibromide

EMAIL THIS PAGE TO A FRIEND
36792 Sigma-Aldrich

1,1′-Diethyl-4,4′-bipyridinium dibromide

puriss., ≥99.0% (AT)

Synonym: Ethyl viologen dibromide

  • CAS Number 53721-12-3

  • Empirical Formula (Hill Notation) C14H18Br2N2

  • Molecular Weight 374.11

  •  MDL number MFCD00134590

Purchase

Properties

grade   puriss.
assay   ≥99.0% (AT)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   275-285 °C (dec.)

Description

Other Notes

Electrochemical properties

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

384097 Ethyl viologen dibromide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?