• USA Home
  • 37862 - Methyl-DOPA sesquihydrate

EMAIL THIS PAGE TO A FRIEND
37862 Sigma-Aldrich

Methyl-DOPA sesquihydrate

≥99.0% (sum of enantiomers, TLC)

Synonym: 3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine, MK-351, Methyl-L-DOPA

  • CAS Number 41372-08-1

  • Empirical Formula (Hill Notation) C10H13NO4 · 1.5H2O

  • Molecular Weight 238.24

  •  Beilstein Registry Number 2417244

  •  EC Number 209-089-2

  •  MDL number MFCD00004186

  •  PubChem Substance ID 24863565

Purchase

Properties

InChI Key   CJCSPKMFHVPWAR-JTQLQIEISA-N
assay   ≥99.0% (sum of enantiomers, TLC)
optical activity   [α]20/D −13.0±1°, c = 1% in H2O
impurities   ≤0.5% alanine
  ~12% water
mp   ≥300 °C

Description

Other Notes

Antibacterial properties

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YP2860000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

857416 (−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?