EMAIL THIS PAGE TO A FRIEND
45318 Sigma-Aldrich

Allidochlor

PESTANAL®, analytical standard

Synonym: N,N-Diallyl-2-chloroacetamide

  • CAS Number 93-71-0

  • Empirical Formula (Hill Notation) C8H12ClNO

  • Molecular Weight 173.64

  •  Beilstein Registry Number 1761616

  •  EC Number 202-270-7

  •  MDL number MFCD00034186

  •  PubChem Substance ID 329756641

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Amide, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   MDBGGTQNNUOQRC-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
2
RTECS 
AB5250000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?