EMAIL THIS PAGE TO A FRIEND
45397 Sigma-Aldrich

Chlorthal-dimethyl

PESTANAL®, analytical standard

Synonym: Dimethyl tetrachloroterephthalate

  • CAS Number 1861-32-1

  • Empirical Formula (Hill Notation) C10H6Cl4O4

  • Molecular Weight 331.96

  •  Beilstein Registry Number 1888840

  •  EC Number 217-464-7

  •  MDL number MFCD00014902

  •  PubChem Substance ID 329756701

Purchase

Properties

Related Categories A-D, Additional Standards, All Herbicide Standards, Alpha Index of All Pesticide Standards, Analytical Standards,
grade   analytical standard
InChI Key   NPOJQCVWMSKXDN-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WZ1500000
Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 3) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?