EMAIL THIS PAGE TO A FRIEND
45486 Sigma-Aldrich

Fenfuram

PESTANAL®, analytical standard

Synonym: 2-Methyl-3-furoic anilide

  • CAS Number 24691-80-3

  • Empirical Formula (Hill Notation) C12H11NO2

  • Molecular Weight 201.22

  •  Beilstein Registry Number 153863

  •  EC Number 246-421-5

  •  MDL number MFCD00055323

  •  PubChem Substance ID 329756778

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Amide structure, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   JFSPBVWPKOEZCB-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
LT8506000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?