EMAIL THIS PAGE TO A FRIEND
45496 Sigma-Aldrich

Ferbam

PESTANAL®, analytical standard

Synonym: Iron(III) dimethyldithiocarbamate

  • CAS Number 14484-64-1

  • Empirical Formula (Hill Notation) C9H18FeN3S6

  • Molecular Weight 416.49

  •  Beilstein Registry Number 3706105

  •  EC Number 238-484-2

  •  MDL number MFCD00067269

  •  PubChem Substance ID 329756788

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Dithiocarbamates, E-G,
grade   analytical standard
description   technical mixture
InChI Key   WHDGWKAJBYRJJL-UHFFFAOYSA-K
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
NO8750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?