EMAIL THIS PAGE TO A FRIEND
45679 Sigma-Aldrich

Tetrachlorvinphos

PESTANAL®, analytical standard

Synonym: Stirofox

  • CAS Number 22248-79-9

  • Empirical Formula (Hill Notation) C10H9Cl4O4P

  • Molecular Weight 365.96

  •  Beilstein Registry Number 1890909

  •  EC Number 244-865-4

  •  MDL number MFCD00078729

  •  PubChem Substance ID 329756893

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Insecticides, Organophorous,
grade   analytical standard
InChI Key   UBCKGWBNUIFUST-YHYXMXQVSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   97-98 °C (lit.)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
TB9100000

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 1) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?