• USA Home
  • 47690 - Formamidine acetate salt

EMAIL THIS PAGE TO A FRIEND
47690 Sigma-Aldrich

Formamidine acetate salt

purum, ≥98.0% (NT)

Synonym: Formamidinium acetate

  • CAS Number 3473-63-0

  • Linear Formula HN=CHNH2 · CH3COOH

  • Molecular Weight 104.11

  •  Beilstein/REAXYS Number 3563488

  •  EC Number 222-442-5

  •  MDL number MFCD00012866

  •  PubChem Substance ID 329757335

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Imines & Amidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
grade   purum
assay   ≥98.0% (NT)
impurities   ≤0.5% formamide
mp   ~160 °C (dec.)
SMILES string   NC=N.CC(O)=O
InChI   1S/C2H4O2.CH4N2/c1-2(3)4;2-1-3/h1H3,(H,3,4);1H,(H3,2,3)
InChI key   XPOLVIIHTDKJRY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?