EMAIL THIS PAGE TO A FRIEND
49670 Sigma-Aldrich

Glutaric anhydride

technical, ≥90% (NT)

  • CAS Number 108-55-4

  • Empirical Formula (Hill Notation) C5H6O3

  • Molecular Weight 114.10

  •  Beilstein/REAXYS Number 110051

  •  EC Number 203-593-6

  •  MDL number MFCD00006679

  •  PubChem Substance ID 329757501

Purchase

Properties

grade   technical
assay   ≥90% (NT)
bp   150 °C/10 mmHg (lit.)
mp   50-55 °C (lit.)
  51-55 °C
SMILES string   O=C1CCCC(=O)O1
InChI   1S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2
InChI key   VANNPISTIUFMLH-UHFFFAOYSA-N

Description

Other Notes

This product has been replaced by G3806-ALDRICH | Glutaric anhydride 95%

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MA3850000
Flash Point(F) 
343.4 °F
Flash Point(C) 
173 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G3806 Glutaric anhydride, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?