• USA Home
  • 50510 - 18β-Glycyrrhetinic acid

EMAIL THIS PAGE TO A FRIEND
50510 Sigma-Aldrich

18β-Glycyrrhetinic acid

purum, ≥97.0% (T)

Synonym: 3β-Hydroxy-11-oxo-18β,20β-olean-12-en-29-oic acid, Enoxolone, Glycyrrhetin, Subglycyrrhelinic acid, Uralenic acid

  • CAS Number 471-53-4

  • Empirical Formula (Hill Notation) C30H46O4

  • Molecular Weight 470.68

  •  Beilstein/REAXYS Number 2229654

  •  EC Number 207-444-6

  •  MDL number MFCD00003706

  •  PubChem Substance ID 329757606

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Complex Molecules
grade   purum
assay   ≥97.0% (T)
optical activity   [α]20/D +165±3°, c = 1% in chloroform
mp   292-295 °C (lit.)
SMILES string   CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(O)=O
InChI   1S/C30H46O4/c1-25(2)21-8-11-30(7)23(28(21,5)10-9-22(25)32)20(31)16-18-19-17-27(4,24(33)34)13-12-26(19,3)14-15-29(18,30)6/h16,19,21-23,32H,8-15,17H2,1-7H3,(H,33,34)/t19-,21-,22-,23+,26+,27-,28-,29+,30+/m0/s1
InChI key   MPDGHEJMBKOTSU-YKLVYJNSSA-N
Gene Information   human ... HSD11B1(3290), HSD11B2(3291)
rat ... Hsd11b1(25116), Hsd11b2(25117)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RK0180000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Steroidal Glycosides on Ascentis® Phenyl
Keywords: Chromatography, High performance liquid chromatography, Vitamins

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G10105 18β-Glycyrrhetinic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?