• USA Home
  • 50719 - Palladium Standard for ICP

EMAIL THIS PAGE TO A FRIEND
50719 Supelco

Palladium Standard for ICP

TraceCERT®, 10,000 mg/L Pd in hydrochloric acid

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application CRMs, Certified Reference Materials (CRMs),
grade   certified reference material
  TraceCERT®
concentration   10,000 mg/L Pd in hydrochloric acid
application(s)   AAS: suitable
  ICP: suitable
format   single component solution

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Preparation Note

10′000 mg/L Pd in 10% HCl (contains HNO3 traces), prepared with high purity Pd metal, HCl, HNO3 and water

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?