EMAIL THIS PAGE TO A FRIEND
51800 Sigma-Aldrich

2-Heptanol

purum, ≥99.0% (GC)

Synonym: Methyl pentyl carbinol

  • CAS Number 543-49-7

  • Linear Formula CH3(CH2)4CH(OH)CH3

  • Molecular Weight 116.20

  •  Beilstein/REAXYS Number 1719088

  •  EC Number 208-844-3

  •  MDL number MFCD00004587

  •  PubChem Substance ID 329757745

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Cell Biology, Chemical Synthesis,
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.420 (lit.)
  n20/D 1.420
bp   160-162 °C (lit.)
density   0.817 g/mL at 25 °C (lit.)
SMILES string   CCCCCC(C)O
InChI   1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3
InChI key   CETWDUZRCINIHU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
RTECS 
MJ2975000
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?