• USA Home
  • 52590 - Hexamethonium bromide

EMAIL THIS PAGE TO A FRIEND
52590 Sigma-Aldrich

Hexamethonium bromide

≥95.0% (AT)

Synonym: Hexane-1,6-bis(trimethylammonium bromide), N,N,N,N′,N′,N′-Hexamethylhexamethylenediammonium dibromide

  • CAS Number 55-97-0

  • Linear Formula (CH3)3N(Br)(CH2)6N(Br)(CH3)3

  • Molecular Weight 362.19

  •  Beilstein/REAXYS Number 3715749

  •  EC Number 200-249-7

  •  MDL number MFCD00011787

  •  PubChem Substance ID 329757858

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
assay   ≥95.0% (AT)
mp   282-285 °C (dec.)
SMILES string   [Br-].[Br-].C[N+](C)(C)CCCCCC[N+](C)(C)C
InChI   1S/C12H30N2.2BrH/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;;/h7-12H2,1-6H3;2*1H/q+2;;/p-2
InChI key   FAPSXSAPXXJTOU-UHFFFAOYSA-L
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
BQ8575000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?