EMAIL THIS PAGE TO A FRIEND
53560 Sigma-Aldrich

Homogentisic acid

purum, ≥99.0% (GC)

Synonym: 2,5-Dihydroxyphenylacetic acid

  • CAS Number 451-13-8

  • Linear Formula (HO)2C6H3CH2CO2H

  • Molecular Weight 168.15

  •  Beilstein Registry Number 2692860

  •  EC Number 207-192-7

  •  MDL number MFCD00004324

  •  PubChem Substance ID 24878091

Purchase

Properties

InChI Key   IGMNYECMUMZDDF-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
mp   150-152 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AH0589000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H0751 Homogentisic acid, crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?