• USA Home
  • 55939 - (S)-3-(4-Chloro-3-fluorophenoxy)-N-(4-cyano-3-trifluoromethylphenyl)-2-hydroxy-2-methylpropionamide

EMAIL THIS PAGE TO A FRIEND
55939 Supelco

(S)-3-(4-Chloro-3-fluorophenoxy)-N-(4-cyano-3-trifluoromethylphenyl)-2-hydroxy-2-methylpropionamide

analytical standard

Synonym: S-23

  • CAS Number 1010396-29-8

  • Empirical Formula (Hill Notation) C18H13ClF4N2O3

  • Molecular Weight 416.75

  •  Beilstein/REAXYS Number 19161994

  •  MDL number MFCD24849232

  •  PubChem Substance ID 329758106

  •  NACRES NA.24

Purchase

Properties

Related Categories Anabolic Agent / SARMS / Other, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol, Steroids / Doping Agents Standards More...
Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
optical activity   [α]/D 20±3°, c = 0.1 in chloroform
shelf life   limited shelf life, expiry date on the label
composition   carbon content, 51.88%
  hydrogen content, 3.14%
  nitrogen content, 6.72%
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   neat
SMILES string   C[C@](O)(COc1ccc(Cl)c(F)c1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
InChI   1S/C18H13ClF4N2O3/c1-17(27,9-28-12-4-5-14(19)15(20)7-12)16(26)25-11-3-2-10(8-24)13(6-11)18(21,22)23/h2-7,27H,9H2,1H3,(H,25,26)/t17-/m0/s1
InChI key   SSFVOEAXHZGTRJ-KRWDZBQOSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?