• USA Home
  • 56458 - (±)-3,4-Methylenedioxyamphetamine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
56458 Supelco

(±)-3,4-Methylenedioxyamphetamine hydrochloride solution

~1 mg/mL in methanol, analytical standard, for drug analysis

  • CAS Number 6292-91-7

  • Empirical Formula (Hill Notation) C10H13NO2 · HCl

  • Molecular Weight 215.68

  •  MDL number MFCD00058557

  •  PubChem Substance ID 329758214

Purchase

Properties

Related Categories Additional Standards, Amphetamine, Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard, for drug analysis
InChI Key   XLAOKZDOZHZWBK-UHFFFAOYSA-N
assay   ≥98.5% (HPLC)
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)
concentration   ~1 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   corresponds for H-NMR
Featured Industry   Pharmaceutical (small molecule)
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

ampoule contains 1.2mL solution

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?