EMAIL THIS PAGE TO A FRIEND
60328 Sigma-Aldrich

Potassium hydride

purum, dispersion (in mineral oil (~35%))

Purchase

Properties

InChI Key   NTTOTNSKUYCDAV-UHFFFAOYSA-N
grade   purum
form   dispersion (in mineral oil (~35%))

Description

Other Notes

Review of its use in organic chemistry

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1409 4.3 / PGI
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

215813 Potassium hydride, 30 wt % dispersion in mineral oil

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?