EMAIL THIS PAGE TO A FRIEND
63005 Sigma-Aldrich

L-(+)-Lyxose

≥99.0% (sum of enantiomers, HPLC)

Synonym: L-Lyxopyranose

  • CAS Number 1949-78-6

  • Empirical Formula (Hill Notation) C5H10O5

  • Molecular Weight 150.13

  •  Beilstein Registry Number 1723082

  •  EC Number 217-763-2

  •  MDL number MFCD00064111

Purchase

Properties

assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +14.0±0.5°, 1 hr, c = 6% in H2O
mp   106-108 °C
  108-112 °C(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?