EMAIL THIS PAGE TO A FRIEND
63010 Sigma-Aldrich

D-(−)-Lyxose

≥99.0% (sum of enantiomers, HPLC)

Synonym: D-Lyxopyranose

  • CAS Number 1114-34-7

  • Empirical Formula (Hill Notation) C5H10O5

  • Molecular Weight 150.13

  •  Beilstein Registry Number 1723083

  •  EC Number 214-212-8

  •  MDL number MFCD00064362

  •  PubChem Substance ID 24882286

Purchase

Properties

assay   ≥99.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −14.0±0.5°, 1 hr, c = 6% in H2O
solubility   H2O: 50 mg/mL, clear, colorless

Description

Other Notes

Use as chiral building block

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

220477 D-(−)-Lyxose, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?