• USA Home
  • 64790 - 2-Methoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
64790 Sigma-Aldrich

2-Methoxybenzoic acid

purum, ≥99.0% (T)

Synonym: O-Methylsalicylic acid, o-Anisic acid

  • CAS Number 579-75-9

  • Linear Formula CH3OC6H4CO2H

  • Molecular Weight 152.15

  •  Beilstein Registry Number 509929

  •  EC Number 209-447-8

  •  MDL number MFCD00002431

  •  PubChem Substance ID 24883555

Purchase

Properties

InChI Key   ILUJQPXNXACGAN-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   98-100 °C(lit.)
  99-102 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?