EMAIL THIS PAGE TO A FRIEND
66712 Sigma-Aldrich

Hexachlorophene

purum, ≥98.0% (T)

Synonym: 2,2′-Methylenebis(3,4,6-trichlorophenol)

  • CAS Number 70-30-4

  • Linear Formula CH2[C6H(Cl)3OH]2

  • Molecular Weight 406.90

  •  Beilstein Registry Number 2064407

  •  EC Number 200-733-8

  •  MDL number MFCD00002171

Purchase

Properties

InChI Key   ACGUYXCXAPNIKK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   161-164 °C
  163-165 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2875 6.1 / PGIII
WGK Germany 
3
RTECS 
SM0700000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?