EMAIL THIS PAGE TO A FRIEND
67750 Sigma-Aldrich

Methylmalonic acid

purum, ≥99.0% (T)

Synonym: 2-Methylpropanedioic acid

  • CAS Number 516-05-2

  • Linear Formula CH3CH(CO2H)2

  • Molecular Weight 118.09

  •  Beilstein Registry Number 1756084

  •  EC Number 208-219-5

  •  MDL number MFCD00002656

  •  PubChem Substance ID 24885379

Purchase

Properties

InChI Key   ZIYVHBGGAOATLY-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   128-130 °C (lit.)
  131-134 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
OO1400000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M54058 Methylmalonic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?