EMAIL THIS PAGE TO A FRIEND
68440 Sigma-Aldrich

4-Methyl-2-pentanol

purum, ≥97.5% (GC)

Synonym: 3-MIC, Isobutyl methyl carbinol, MAOH, MIBC

  • CAS Number 108-11-2

  • Linear Formula (CH3)2CHCH2CH(OH)CH3

  • Molecular Weight 102.17

  •  Beilstein/REAXYS Number 1098268

  •  EC Number 203-551-7

  •  MDL number MFCD00004550

  •  PubChem Substance ID 329760908

Purchase

Properties

vapor density   3.5 (vs air)
vapor pressure   3.7 mmHg ( 20 °C)
grade   purum
assay   ≥97.5% (GC)
expl. lim.   5.5 %
refractive index   n20/D 1.410 (lit.)
  n20/D 1.411
bp   132 °C (lit.)
mp   −90 °C (lit.)
density   0.802 g/mL at 25 °C (lit.)
SMILES string   CC(C)CC(C)O
InChI   1S/C6H14O/c1-5(2)4-6(3)7/h5-7H,4H2,1-3H3
InChI key   WVYWICLMDOOCFB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2053 3 / PGIII
WGK Germany 
1
RTECS 
SA7350000
Flash Point(F) 
105.8 °F
Flash Point(C) 
41 °C
Protocols & Articles

Articles

GC Analysis of C3-C5 Alcohols on Supel-Q™ PLOT

From our library of Articles, Sigma-Aldrich presents GC Analysis of C3-C5 Alcohols on Supel-Q™ PLOT
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

109916 4-Methyl-2-pentanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?