• USA Home
  • 69516 - 4-Methyltrityl chloride polymer-bound

EMAIL THIS PAGE TO A FRIEND
69516 Sigma-Aldrich

4-Methyltrityl chloride polymer-bound

extent of labeling: ~0.7 mmol/g Cl loading

Synonym: 4-Methyltriphenylchloromethane polymer-bound

Purchase

Properties

extent of labeling   ~0.7 mmol/g Cl loading
matrix   polystyrene, crosslinked with 1% DVB
particle size   200-400 mesh

Description

Other Notes

Resin for solid phase peptide synthesis and combinatorial chemistry

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Trityl Resins

Trityl chloride resins and as 4-methyl- and 4-methoxytrityl derivatives render increased acid lability, making them less favorable for stable attachment of C-terminal carboxylic acid during peptide s...
ChemFiles Volume 3 Article 4
Keywords: Immobilization, Peptide synthesis, Solid phase peptide synthesis, Solid supported organic synthesis

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?