• USA Home
  • 70910 - 2-Naphthaleneacetic acid

EMAIL THIS PAGE TO A FRIEND
70910 Sigma-Aldrich

2-Naphthaleneacetic acid

purum, ≥98.0% (T)

Synonym: 2-Naphthylacetic acid

Purchase

Properties

grade   purum
assay   ≥98.0% (T)
ign. residue   ≤0.1%
mp   139-142 °C
  141-143 °C (lit.)
solubility   acetone: 50 mg/mL, clear
SMILES string   OC(=O)Cc1ccc2ccccc2c1
InChI   1S/C12H10O2/c13-12(14)8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7H,8H2,(H,13,14)
InChI key   VIBOGIYPPWLDTI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
QJ0876000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?