EMAIL THIS PAGE TO A FRIEND
71431 Sigma-Aldrich

Sodium cyanide

puriss. p.a., ACS reagent, ≥97.0% (AT)

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, Puriss p.a. ACS,
grade   ACS reagent
vapor density   1.7 (vs air)
vapor pressure   1 mmHg ( 817 °C)
InChI Key   RTVFYQXEHKQMKO-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥97.0% (AT)
mp   563.7 °C(lit.)
solubility   water: soluble(lit.)
anion traces   chloride (Cl-): ≤1500 mg/kg
  phosphate (PO43-): ≤200 mg/kg
  sulfate (SO42-): ≤500 mg/kg
  sulfide (S2-): ≤10 mg/kg
  thiocyanate (SCN-): ≤100 mg/kg
cation traces   Ag: ≤5 mg/kg
  Al: ≤5 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤50 mg/kg
  K: ≤2000 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Tl: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Reagent for the decomposition of cassiterite by fusion

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Sodium cyanide may be employed for the synthesis of the following compounds:
• α-amino acids
• Prussian blue (cyanide pigment)
• nitriles, via reaction with primary and secondary chlorides

General description

Sodium cyanide is colorless sodium salt and its crystals belong to the cubic crystal system. Its aqueous solutions are strongly basic due to the presence of cyanide ion. It can be synthesized by reacting sodium hydroxide with hydrogen cyanide. It is a key precursor for the synthesis of various organic intermediates. It participates in the RuCl3-mediated oxidative cyanation of tertiary amines.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 1689 6.1 / PGI
WGK Germany 
3
RTECS 
VZ7525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?