• USA Home
  • 72372 - N,N-Diethylnicotinamide

EMAIL THIS PAGE TO A FRIEND
72372 Sigma-Aldrich

N,N-Diethylnicotinamide

tested according to Ph Eur

Synonym: ‘Nikethamide’, Nicethamidum, Nicotinic acid diethylamide

  • CAS Number 59-26-7

  • Empirical Formula (Hill Notation) C10H14N2O

  • Molecular Weight 178.23

  •  Beilstein/REAXYS Number 5743

  •  EC Number 200-418-5

  •  MDL number MFCD00006386

  •  PubChem Substance ID 329764029

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
InChI Key   NCYVXEGFNDZQCU-UHFFFAOYSA-N
refractive index   n20/D 1.524 (lit.)
bp   296-300 °C (lit.)
mp   23 °C (lit.)
density   1.06 g/mL at 25 °C (lit.)
Agency/Method   tested according to Ph Eur
Featured Industry   Pharmaceutical (small molecule)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
QS4375000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?