EMAIL THIS PAGE TO A FRIEND
73379 Supelco

Podophyllotoxin

analytical standard

  • CAS Number 518-28-5

  • Empirical Formula (Hill Notation) C22H22O8

  • Molecular Weight 414.41

  •  Beilstein/REAXYS Number 99163

  •  EC Number 208-250-4

  •  MDL number MFCD00075290

  •  PubChem Substance ID 329764848

  •  NACRES NA.24

Purchase

Properties

Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
optical activity   [α]/D 131±2°, c = 1 in chloroform
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   183-184 °C (lit.)
format   neat
storage temp.   2-8°C
SMILES string   COc1cc(cc(OC)c1OC)[C@H]2C3C(COC3=O)[C@@H](O)c4cc5OCOc5cc24
InChI   1S/C22H22O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,13,18-20,23H,8-9H2,1-3H3/t13-,18+,19-,20-/m0/s1
InChI key   YJGVMLPVUAXIQN-XVVDYKMHSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Inhibits microtubule assembly; antineoplastic.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
UN 3462 6.1 / PGII
WGK Germany 
WGK 3
RTECS 
LV2500000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?