EMAIL THIS PAGE TO A FRIEND
74090 Sigma-Aldrich

3-Nitro-L-tyrosine

purum, ≥98.0% (NT)

  • CAS Number 621-44-3

  • Linear Formula O2NC6H3-4-(OH)CH2CH(NH2)CO2H

  • Molecular Weight 226.19

  •  Beilstein Registry Number 2813157

  •  EC Number 210-688-6

  •  MDL number MFCD00007123

  •  PubChem Substance ID 24886641

Purchase

Properties

InChI Key   FBTSQILOGYXGMD-LURJTMIESA-N
grade   purum
assay   ≥98.0% (NT)
optical activity   [α]20/D +4.5±0.5°, c = 3.2% in 1 M HCl
mp   ~230 °C (dec.)
  233-235 °C (dec.)(lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?