EMAIL THIS PAGE TO A FRIEND
75152 Supelco

3-Methylpiperidine

analytical standard

Synonym: β-Pipecoline, 3-Pipecoline

  • CAS Number 626-56-2

  • Empirical Formula (Hill Notation) C6H13N

  • Molecular Weight 99.17

  •  Beilstein/REAXYS Number 79807

  •  EC Number 210-953-6

  •  MDL number MFCD00005994

  •  PubChem Substance ID 329766223

Purchase

Properties

Quality Level   100
grade   analytical standard
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.447 (lit.)
bp   125-126 °C/763 mmHg (lit.)
density   0.845 g/mL at 25 °C (lit.)
storage temp.   room temp
SMILES string   CC1CCCNC1
InChI   1S/C6H13N/c1-6-3-2-4-7-5-6/h6-7H,2-5H2,1H3
InChI key   JEGMWWXJUXDNJN-UHFFFAOYSA-N

Description

Application

Reactant for synthesis of:
• Phenylpropenamide derivatives for anti-hepatitis B virus activity
• CB2 receptor agonists for treatment of chronic pain
• Aurora kinase inhibitors
• Spiroimidazolidinone NPC1L1 inhibitors
• 1,3,5-Oxadiazinones
• Selective serotonin 5-HT6 receptor antagonists

Safety & Documentation

Safety Information

RIDADR 
UN 1993BF 3 / PGII
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?