EMAIL THIS PAGE TO A FRIEND
76710 Sigma-Aldrich

Pentafluorobenzene

purum, ≥99.0% (GC)

Purchase

Properties

InChI Key   WACNXHCZHTVBJM-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.391(lit.)
  n20/D 1.391
bp   85 °C(lit.)
mp   −48 °C(lit.)
density   1.514 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
RTECS 
DA6651600
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?