EMAIL THIS PAGE TO A FRIEND
76940 Sigma-Aldrich

(R)-(−)-2-Pentanol

purum, ≥98.0% (sum of enantiomers, GC)

  • CAS Number 31087-44-2

  • Linear Formula CH3CH2CH2CH(OH)CH3

  • Molecular Weight 88.15

  •  Beilstein/REAXYS Number 4652311

  •  MDL number MFCD00065953

  •  PubChem Substance ID 329767441

Purchase

Properties

vapor density   3 (vs air)
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −13±1°, neat
optical purity   enantiomeric ratio: ≥95:5 (GC)
autoignition temp.   657 °F
expl. lim.   9 %
refractive index   n20/D 1.406 (lit.)
  n20/D 1.407
bp   119-120 °C (lit.)
density   0.809 g/mL at 20 °C (lit.)
SMILES string   CCC[C@@H](C)O
InChI   1S/C5H12O/c1-3-4-5(2)6/h5-6H,3-4H2,1-2H3/t5-/m1/s1
InChI key   JYVLIDXNZAXMDK-RXMQYKEDSA-N

Description

Other Notes

This chiral alcohol and its derivatives are used as reference substances in many asymmetric reactions, e.g.; Synthesis of the stereochemically pure bromide

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1105 - class 3 - PG 3 - Pentanols
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

330523 (R)-(−)-2-Pentanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?