• USA Home
  • 77376 - (±)-Metoprolol (+)-tartrate salt

EMAIL THIS PAGE TO A FRIEND
77376 Supelco

(±)-Metoprolol (+)-tartrate salt

pharmaceutical secondary standard; traceable to USP, PhEur and BP

Synonym: (±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor

Purchase

Properties

InChI Key   YGULWPYYGQCFMP-CEAXSRTFSA-N
pharmacopeia traceability   traceable to BP 540
  traceable to PhEur M1830000
  traceable to USP 1441301
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UB7450100
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?