• USA Home
  • 77670 - 4-Hydroxybenzenesulfonic acid solution

EMAIL THIS PAGE TO A FRIEND
77670 Sigma-Aldrich

4-Hydroxybenzenesulfonic acid solution

technical, ~65% in H2O

Synonym: Phenol-4-sulfonic acid

  • CAS Number 98-67-9

  • Linear Formula HOC6H4SO3H

  • Molecular Weight 174.17

  •  Beilstein Registry Number 1869034

  •  MDL number MFCD00007506

  •  PubChem Substance ID 24887214

Purchase

Properties

InChI Key   FEPBITJSIHRMRT-UHFFFAOYSA-N
grade   technical
concentration   ~65% in H2O
impurities   ≤2% H2SO4
  ≤2% phenol
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2586PSN2 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?