• USA Home
  • 79763 - (2RS)-2-(4-Isobutylphenyl)propionamide

EMAIL THIS PAGE TO A FRIEND
79763 Supelco

(2RS)-2-(4-Isobutylphenyl)propionamide

pharmaceutical impurity standard

Synonym: α-Methyl-4-(2-methylpropyl)benzeneacetamide, (±)-Ibuprofen amide, (2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide, Ibuprofen impurity C (PhEur)

  • CAS Number 59512-17-3

  • Empirical Formula (Hill Notation) C13H19NO

  • Molecular Weight 205.30

  •  Beilstein/REAXYS Number 2448195

  •  EC Number 261-793-9

  •  MDL number MFCD00454181

  •  PubChem Substance ID 329768615

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Ibuprofen Impurities, Pharmaceutical Impurities, Pharmacopeia & Metrological Institutes Standards More...
Quality Level   100
InChI Key   REUQKCDCQVNKLW-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
shelf life   limited shelf life, expiry date on the label
impurities   ≤2.0% related substances (HPLC)
  ≤2.0% water (calc. from elemental analysis)
mp   112-118 °C
Featured Industry   Pharmaceutical (small molecule)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?