EMAIL THIS PAGE TO A FRIEND
79810 Sigma-Aldrich

Phthalocyanine

for fluorescence

  • CAS Number 574-93-6

  • Empirical Formula (Hill Notation) C32H18N8

  • Molecular Weight 514.54

  •  Beilstein Registry Number 378323

  •  EC Number 209-378-3

  •  MDL number MFCD00005085

Purchase

Properties

grade   for fluorescence
fluorescence   λex 350 nm; λem 700 nm in 0.1 M citrate pH 2.0
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?