EMAIL THIS PAGE TO A FRIEND
80860 Sigma-Aldrich

Piperonylic acid

purum, ≥97.0% (T)

Synonym: 1,3-Benzodioxole-5-carboxylic acid, 3,4-(Methylenedioxy)benzoic acid

  • CAS Number 94-53-1

  • Empirical Formula (Hill Notation) C8H6O4

  • Molecular Weight 166.13

  •  Beilstein/REAXYS Number 150206

  •  EC Number 202-342-8

  •  MDL number MFCD00005830

  •  PubChem Substance ID 329769112

Purchase

Properties

grade   purum
assay   ≥97.0% (T)
mp   229-231 °C (lit.)
  230-232 °C
SMILES string   OC(=O)c1ccc2OCOc2c1
InChI   1S/C8H6O4/c9-8(10)5-1-2-6-7(3-5)12-4-11-6/h1-3H,4H2,(H,9,10)
InChI key   VDVJGIYXDVPQLP-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DF4912765

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?