EMAIL THIS PAGE TO A FRIEND
81125 Sigma-Aldrich

Podophyllotoxin

≥98.0% (HPLC)

  • CAS Number 518-28-5

  • Empirical Formula (Hill Notation) C22H22O8

  • Molecular Weight 414.41

  •  Beilstein Registry Number 99163

  •  EC Number 208-250-4

  •  MDL number MFCD00075290

  •  PubChem Substance ID 24887649

Purchase

Properties

InChI Key   YJGVMLPVUAXIQN-XVVDYKMHSA-N
assay   ≥98.0% (HPLC)
optical activity   [α]20/D −132±4°, c = 1% in chloroform
mp   183-184 °C(lit.)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Inhibits microtubule assembly; antineoplastic.

Other Notes

Inhibitor of microtubule assembly. Useful tool for tubulin studies

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3462 6.1 / PGII
WGK Germany 
3
RTECS 
LV2500000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?