• USA Home
  • 81515 - Prazosin hydrochloride

EMAIL THIS PAGE TO A FRIEND
81515 Sigma-Aldrich

Prazosin hydrochloride

≥99.0% (TLC)

Synonym: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine hydrochloride, Furazosin hydrochloride

  • CAS Number 19237-84-4

  • Empirical Formula (Hill Notation) C19H21N5O4 · HCl

  • Molecular Weight 419.86

  •  Beilstein Registry Number 4303561

  •  EC Number 242-903-4

  •  MDL number MFCD00058177

Purchase

Properties

InChI Key   WFXFYZULCQKPIP-UHFFFAOYSA-N
assay   ≥99.0% (TLC)
impurities   ≤1% water
solubility   H2O: 1 mg/mL, clear, colorless

Description

Other Notes

An α1-adrenergic receptor antagonist. Suppresses experimental autoimmune encephalomyelitis in the Lewis rat

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VA1350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P7791 Prazosin hydrochloride, ≥99.0% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?