• USA Home
  • 81664 - Procainamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
81664 Sigma-Aldrich

Procainamide hydrochloride

purum, ≥98.0% (AT)

Synonym: 4-Amino-N-(2-diethylaminoethyl)benzamide hydrochloride, 4-Aminobenzoic acid 2-diethylaminoethylamide

  • CAS Number 614-39-1

  • Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl

  • Molecular Weight 271.79

  •  Beilstein Registry Number 3729517

  •  EC Number 210-381-7

  •  MDL number MFCD00012998

  •  PubChem Substance ID 329769213

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   ABTXGJFUQRCPNH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   165-168 °C
  167-169 °C (lit.)
solubility   H2O: 1 g/10 mL, clear, greenish-yellow
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CV2295000

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?