EMAIL THIS PAGE TO A FRIEND
84220 Sigma-Aldrich

Ethyl salicylate

purum, ≥99.0% (GC)

Synonym: Ethyl 2-hydroxybenzoate

  • CAS Number 118-61-6

  • Linear Formula 2-(HO)C6H4CO2C2H5

  • Molecular Weight 166.17

  •  Beilstein Registry Number 907659

  •  EC Number 204-265-5

  •  MDL number MFCD00002215

  •  PubChem Substance ID 24888187

Purchase

Properties

vapor pressure   0.05 mmHg ( 25 °C)
InChI Key   GYCKQBWUSACYIF-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.522(lit.)
  n20/D 1.523
bp   234 °C(lit.)
mp   1 °C (lit.)
density   1.131 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
VO3000000
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

112291 Ethyl salicylate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?