• USA Home
  • 85036 - Serotonin hydrochloride

EMAIL THIS PAGE TO A FRIEND
85036 Sigma-Aldrich

Serotonin hydrochloride

≥98.0% (HPLC)

Synonym: 3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride

  • CAS Number 153-98-0

  • Empirical Formula (Hill Notation) C10H12N2O · HCl

  • Molecular Weight 212.68

  •  Beilstein/REAXYS Number 3570963

  •  MDL number MFCD00012686

Purchase

Properties

assay   ≥98.0% (HPLC)
impurities   ≤5% ethanol
  ≤5% water
mp   149-154 °C (lit.)
  177-180 °C
SMILES string   Cl[H].NCCc1c[nH]c2ccc(O)cc12
InChI   1S/C10H12N2O.ClH/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;/h1-2,5-6,12-13H,3-4,11H2;1H
InChI key   MDIGAZPGKJFIAH-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Neurotransmitter.

Other Notes

Reviews on: the physiological activities of serotonin; serotonin neurochemistry; the mechanisms of serotonergic signal transduction.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM2571000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

H9523 Serotonin hydrochloride, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?