EMAIL THIS PAGE TO A FRIEND
85472 Sigma-Aldrich

α-Solanine

from potato sprouts, ≥99.0% (TLC)

Synonym: alpha-Solanine

  • CAS Number 20562-02-1

  • Empirical Formula (Hill Notation) C45H73NO15

  • Molecular Weight 868.06

  •  Beilstein Registry Number 77607

  •  EC Number 243-879-8

  •  MDL number MFCD00077873

  •  PubChem Substance ID 24888360

Purchase

Properties

biological source   potato sprouts
InChI Key   ZGVSETXHNHBTRK-UDJLNJFBSA-N
assay   ≥99.0% (TLC)
storage temp.   −20°C

Description

Other Notes

Insecticide properties

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WF0250000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S3757 α-Solanine, from potato sprouts, ≥95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?