EMAIL THIS PAGE TO A FRIEND
85882 Sigma-Aldrich

Streptozocin

≥98.0% (HPLC)

Synonym: N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozotocin

  • CAS Number 18883-66-4

  • Empirical Formula (Hill Notation) C8H15N3O7

  • Molecular Weight 265.22

  •  Beilstein Registry Number 2060675

  •  EC Number 242-646-8

  •  MDL number MFCD00006607

  •  PubChem Substance ID 329769421

Purchase

Properties

InChI Key   ZSJLQEPLLKMAKR-GKHCUFPYSA-N
assay   ≥98.0% (HPLC)
mp   121 °C (dec.)(lit.)
Mode of action   DNA synthesis | interferes
storage temp.   −20°C

Description

Biochem/physiol Actions

An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets; induces death of insulin-secreting cells, producing an animal model of diabetes. Potent DNA methylating agent that induces chromosomal breakage. Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter.

General description

Chemical structure: aminoglycoside

Other Notes

This product has been replaced by S0130-SIGMA | Streptozocin ≥75% α-anomer basis, ≥98% (HPLC), powder

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
LZ5775000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S0130 Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?