• USA Home
  • 88002 - 1,1,2,2-Tetraphenyl-1,2-ethanediol

EMAIL THIS PAGE TO A FRIEND
88002 SAFC

1,1,2,2-Tetraphenyl-1,2-ethanediol

purum, ≥98.0% (HPLC)

Synonym: Benzopinacol

  • CAS Number 464-72-2

  • Linear Formula (C6H5)2C(OH)C(C6H5)2OH

  • Molecular Weight 366.45

  •  Beilstein Registry Number 2061947

  •  EC Number 207-356-8

  •  MDL number MFCD00004448

  •  PubChem Substance ID 24889034

Purchase

Properties

InChI Key   MFEWNFVBWPABCX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   171-173 °C (lit.)
  171-175 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
KI2750000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?