• USA Home
  • 88126 - Tetrapropylammonium perruthenate

EMAIL THIS PAGE TO A FRIEND
88126 Sigma-Aldrich

Tetrapropylammonium perruthenate

purum

Synonym: TPAP

Purchase

Properties

InChI Key   NQSIKKSFBQCBSI-UHFFFAOYSA-N
grade   purum
mp   ~160 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Catalyst for the mild N-methylmorpholine N-oxide, NMO, mediated oxidation of alcohols to carbonyl compounds in methylene chloride

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?